Stampa…

Tristearin (glycerol tristearate) ≥80.0% (by GC)

Fornitore: TCI

Synonyms: Propane-1,2,3-triyl tristearate, Tristearin

G0212-250G G0212-25G
TCIAG0212-250GEA 159 EUR
TCIAG0212-250G TCIAG0212-25G
Tristearin (glycerol tristearate) ≥80.0% (by GC)
Tristearin (glycerol tristearate)
Formula: C₅₇H₁₁₀O₆
Peso molecolare: 891.4797 g/mol
MDL Number: MFCD00036230
CAS Number: 555-43-1
EINECS: 209-097-6
Merck Index: 14,09756
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=337c43d5901254c397bbadb82b4f6d27b35932a5&vid=337c43d5901254c397bbadb82b4f6d27b35932a5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 80.0 %
Melting point 66.0 to 74.0 deg-C
Solubility in hot Toluene almost transparency


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR