2-Methylbenzamidoxime
Fornitore: Cayman Chemical
13002-500
13002-1
13002-5
CAYM13002-500EA
68.5
EUR
CAYM13002-500
CAYM13002-1
CAYM13002-5
2-Methylbenzamidoxime
2-Methylbenzamidoxime
Formula:
C₈H₁₀N₂O Peso molecolare: 150,18 g/mol |
CAS Number:
40312-14-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...