Stampa…
Flasks, round bottom, narrow neck, with beaded rim
Fornitore: witeg Labortechnik
8.609.025 8.609.100 8.609.250
WITG8.609.025EA 0 EUR
WITG8.609.025 WITG8.609.100 WITG8.609.250
Flasks, round bottom, narrow neck, with beaded rim
Fiasche Palloni a fondo tondo
Quartz glass, clear.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Marked with capacity and a white labelling area.