1-Chloro-4-phenylphthalazine 96%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 96%
CAS No.: 10132-01-1
Molecular Formula: C14H9ClN2
Formula Weight: 240.69
Melting Point: 158-159°
MDL No.: MFCD00524786
Formula:
C₁₄H₉ClN₂ Punto di fusione: 158…159 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00524786 CAS Number: 10132-01-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...