N,N-Diphenylcarbamoyl chloride 98%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
167140250
167141000
167145000
ACRO167140250EA
42.8
EUR
ACRO167140250
ACRO167141000
ACRO167145000
N,N-Diphenylcarbamoyl chloride 98%
N,N-Diphenylcarbamoyl chloride
Formula:
C₁₃H₁₀ClNO Punto di fusione: 81…85 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00000633 CAS Number: 83-01-2 EINECS: 201-450-2 UN: 3261 ADR: 8,II |
Risultati del test di specificazione
Appearance | Beige to grey-green Crystalline powder |
Infrared spectrum | Conforms |
Melting point | 81 to 85 °C |
GC | ≥97.5 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...