Stampa…

L-Methionine methyl ester hydrochloride ≥99%

Fornitore: Thermo Scientific

Synonyms: H-Met-OMe.HCl, H-Met-OMe·HCl

A15952.06 A15952.14 A15952.22
A15952.06EA 36.1 EUR
A15952.06 A15952.14 A15952.22
L-Methionine methyl ester hydrochloride ≥99%
L-Methionine methyl ester hydrochloride

Purity: 99% CAS No.: 2491-18-1 Molecular Formula: C6H13NO2S.HCl Formula Weight: 199.70 Storage Temperature: -30°C to -10°C Melting Point: 151-152° Beilstein Reference: 3913214 Optical Rotation: +26° (c=5 in water) MDL No.: MFCD00012491

Formula: C₆H₁₃NO₂S·HCl
Peso molecolare: 199,7 g/mol
Punto di fusione: 151…152 °C
MDL Number: MFCD00012491
CAS Number: 2491-18-1
EINECS: 219-651-9

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR