Stampa…
Cumene hydroperoxide ≥80% in cumene per sintesi, Sigma-Aldrich®
Fornitore: Merck
Pericolo
Synonyms: Cumyl hydroperoxide
M-Clarity™ quality level = MQ200
8.20502.0005 8.20502.0250
8.20502.0005EA 19.8 EUR
8.20502.0005 8.20502.0250
Cumene hydroperoxide ≥80% in cumene per sintesi, Sigma-Aldrich®
Cumene hydroperoxide
Formula: C₉H₁₂O₂
Peso molecolare: 152,20 g/mol
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00002129
CAS Number: 80-15-9
EINECS: 201-254-7
UN: 3109
ADR: 5.2 (8),
Visualizza i codici
Solubility 13 g/l (20 °C)
Boiling Point 116 °C (20 hPa)
Flash Point 57 °C
Refractive Index 1.5242 (20 °C, 589 nm)
Density 1.03 g/cm3 (20 °C)
pH 5 (13 g/l, H2O, 20 °C)
Synonyms α,α-Dimethylbenzyl hydroperoxide