Stampa…

Dioctyl phenylphosphonate ≥97%

Fornitore: Thermo Scientific

Synonyms: Di-n-octyl phenylphosphonate

A13468.06 A13468.14 A13468.22
A13468.06EA 40.7 EUR
A13468.06 A13468.14 A13468.22
Dioctyl phenylphosphonate ≥97%
Dioctyl phenylphosphonate

Purity: 97% CAS No.: 1754-47-8 Molecular Formula: C22H39O3P Formula Weight: 382.53 Flash Point: 110°(230°F) Refractive Index: 1.4795 Boiling Point: 219-220°/0.7mm Beilstein Reference: 2948496 MDL No.: MFCD00009561

Formula: C₂₂H₃₉O₃P
Peso molecolare: 382,53 g/mol
Punto di ebollizione: 219…220 °C (0,7 mmHg)
Densità: 0,932
Flash Pt: >110 °C (230 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00009561
CAS Number: 1754-47-8
EINECS: 217-142-6

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR