(Methylthio)acetone ≥98%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 98+%
CAS No.: 14109-72-9
Molecular Formula: C4H8OS
Formula Weight: 104.17
Flash Point: 43°(109°F)
Refractive Index: 1.4740
Odour: Stench
Boiling Point: 69-70°/30mm
Beilstein Reference: 1737738
UN No.: 1224
MDL No.: MFCD00015325
Formula:
C₄H₈OS Punto di ebollizione: 69…70 °C (30 mmHg) Densità: 1,023 Flash Pt: 43 °C (109 °F) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00015325 CAS Number: 14109-72-9 UN: 1224 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...