Stampa…

α,α-Difluorophenylacetic acid ≥99%

Fornitore: Thermo Scientific
Pericolo

Synonyms: α,α-Difluorobenzeneacetic acid

H26028.MD H26028.03
H26028.MDEA 56.7 EUR
H26028.MD H26028.03
α,α-Difluorophenylacetic acid ≥99%
α,α-Difluorophenylacetic acid

Purity: 99% CAS No.: 360-03-2 Molecular Formula: C8H6F2O2 Formula Weight: 172.13 Melting Point: 73-76° UN No.: 3261 MDL No.: MFCD00498146

Formula: C₈H₆F₂O₂
Peso molecolare: 172,13 g/mol
Punto di fusione: 73…76 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00498146
CAS Number: 360-03-2
UN: 3261
ADR: 8,III

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR