Dipropyl phthalate ≥98.0% (by GC)
Fornitore: TCI
|
Attenzione
|
P0306-25ML
P0306-500ML
P0306-100ML
TCIAP0306-25MLEA
51
EUR
TCIAP0306-25ML
TCIAP0306-500ML
TCIAP0306-100ML
Dipropyl phthalate ≥98.0% (by GC)
Dipropyl phthalate
Formula:
C₁₄H₁₈O₄ Punto di ebollizione: 305°C |
MDL Number:
MFCD00009371 CAS Number: 131-16-8 EINECS: 205-015-8 UN: 3082 ADR: 9,III |
|
Risultati del test di specificazione
Appearance | Colorless to Light yellow clear liquid |
Purity (GC) | min. 98.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...