Stampa…

Dihexyl phthalate ≥98.0% (by GC)

Fornitore: TCI EUROPE
Pericolo

P1344-25G P1344-5G
TCIAP1344-25GEA 92 EUR
TCIAP1344-25G TCIAP1344-5G
Dihexyl phthalate ≥98.0% (by GC)
Dihexyl phthalate
Formula: C₂₀H₃₀O₄
Punto di ebollizione: 333°C
Flash Pt: 177°C
MDL Number: MFCD00043720
CAS Number: 84-75-3
EINECS: 201-559-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3f13aa1c535f7f563505214464215f86d14b13f8&vid=3f13aa1c535f7f563505214464215f86d14b13f8&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR