Stampa…

Methyl nitroacetate ≥97.0% (by GC)

Fornitore: TCI
Attenzione

N0656-25G N0656-5G
TCIAN0656-25GEA 308 EUR
TCIAN0656-25G TCIAN0656-5G
Methyl nitroacetate ≥97.0% (by GC)
Methyl nitroacetate
Formula: C₃H₅NO₄
Punto di ebollizione: 100°C
Flash Pt: 95°C
MDL Number: MFCD00075480
CAS Number: 2483-57-0
EINECS: 219-622-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6eff02daee5c92c2679d1c5c5b31ffcb945efc4e&vid=6eff02daee5c92c2679d1c5c5b31ffcb945efc4e&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Light yellow clear liquid
Purity (GC) min. 97.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR