5-Bromo-2-chloropyrimidine ≥97.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
B2847-1G
B2847-25G
B2847-5G
TCIAB2847-1GEA
16
EUR
TCIAB2847-1G
TCIAB2847-25G
TCIAB2847-5G
5-Bromo-2-chloropyrimidine ≥97.0% (by GC)
5-Bromo-2-chloropyrimidine
Formula:
C₄H₂BrClN₂ Peso molecolare: 193,43 g/mol Punto di ebollizione: 95°C |
MDL Number:
MFCD00483232 CAS Number: 32779-36-5 |
Risultati del test di specificazione
Appearance | White to Orange to Green powder to crystal |
Purity (GC) | min. 97.0 % |
Melting point | 76.0 to 80.0 deg-C |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...