Stampa…

6-Chlorouracil ≥98.0% (by titrimetric analysis)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 6-Chloro-2,4-dihydroxypyrimidine, 6-Chloropyrimidine-2,4(1H,3H)-dione, 4-Chlorouracil, 4-(3-Bromophenyl)pyrazole, 4-Chloro-2,6-dihydroxypyrimidine, 6-Chloro-2,4(1H,3H)-pyrimidinedione

C2093-25G C2093-5G
TCIAC2093-25GEA 192 EUR
TCIAC2093-25G TCIAC2093-5G
6-Chlorouracil ≥98.0% (by titrimetric analysis)
6-Chlorouracil
Formula: C₄H₃ClN₂O₂
Peso molecolare: 146,53 g/mol
Punto di fusione: 295°C
MDL Number: MFCD00014595
CAS Number: 4270-27-3
EINECS: 224-258-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=9a8f7707165dc78c0d16e81d93d2537748943eca&vid=9a8f7707165dc78c0d16e81d93d2537748943eca&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR