Stampa…

Trimethylsilylacetylene ≥98.0% (by GC)

Fornitore: TCI
Pericolo

T1239-250ML T1239-25ML T1239-5ML
TCIAT1239-250MLEA 390 EUR
TCIAT1239-250ML TCIAT1239-25ML TCIAT1239-5ML
Trimethylsilylacetylene ≥98.0% (by GC)
Trimethylsilylacetylene
Formula: C₅H₁₀Si
Punto di ebollizione: 53°C
Densità: 0,7111 g/cm³ (20 °C)
Flash Pt: -34°C
MDL Number: MFCD00008569
CAS Number: 1066-54-2
EINECS: 213-919-9
UN: 1993
ADR: 3,II
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=55f31012b010cd69de1c6ce9bf6e3f9851cac1e1&vid=55f31012b010cd69de1c6ce9bf6e3f9851cac1e1&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR