Stampa…

3,5-Difluorophenylboronic acid (contains varying amounts of Anhydride)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (3,5-Difluorophenyl)metaboric acid, Dihydroxy(3,5-difluorophenyl)borane, 3,5-Difluorobenzeneboronic acid

D3081-25G D3081-5G
TCIAD3081-25GEA 65 EUR
TCIAD3081-25G TCIAD3081-5G
3,5-Difluorophenylboronic acid (contains varying amounts of Anhydride)
3,5-Difluorophenylboronic acid
Formula: C₆H₅BF₂O₂
Peso molecolare: 157,91 g/mol
Punto di fusione: 250°C
MDL Number: MFCD01318138
CAS Number: 156545-07-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a41d9abb6abbfb19c1e317db5ac3679e927d48d5&vid=a41d9abb6abbfb19c1e317db5ac3679e927d48d5&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance White to Orange to Green powder to crystal
Purity (Neutralization titration) 95.0 to 115.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR