3-Methylxanthine ≥98.0% (by HPLC, titration analysis)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
3,7-Dihydro-3-methyl-1H-purine-2,6-dione, 2,6-Dihydroxy-3-methylpurine
M2073-25G
M2073-5G
TCIAM2073-25GEA
171
EUR
TCIAM2073-25G
TCIAM2073-5G
3-Methylxanthine ≥98.0% (by HPLC, titration analysis)
3-Methylxanthine
Formula:
C₆H₆N₄O₂ Peso molecolare: 166,14 g/mol Punto di fusione: 360°C |
MDL Number:
MFCD00005580 CAS Number: 1076-22-8 EINECS: 214-058-1 |
Risultati del test di specificazione
Appearance | White to Light yellow powder to crystal |
Purity (HPLC) | min. 98.0 area% |
Purity (Neutralization titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...