Tetrabutylammonium tribromide ≥98.0% (by titrimetric analysis)
Fornitore: TCI
|
Pericolo
|
Synonyms:
Tetra-n-butylammonium tribromide, N,N,N,N-Tetrabutylammonium tribromide
Formula:
C₁₆H₃₆Br₃N Punto di fusione: 76 °C |
MDL Number:
MFCD00012110 CAS Number: 38932-80-8 UN: 1759 ADR: 8,III |
|
Risultati del test di specificazione
Appearance | White to Yellow to Orange powder to crystal |
Purity (Iodometric Titration) | min. 98.0 % |
Melting point | 73.0 to 77.0 deg-C |
Solubility in hot Methanol | almost transparency |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...