Stampa…
Storage containers, Brute®
Fornitore: Rubbermaid Commercial Products
FG262000YEL FG262000BLUE
129-1857EA 107 EUR
129-1857 129-1861
Storage containers, Brute®
Scatole Scatole di stoccaggio
These containers with stackable design enables nesting for easy transport and storage making refuse collection and storage more efficient.
  • Innovative venting channels make lifting out liners easier improving productivity and reducing the risk of injury
  • Integrated cinches secure the liner, allowing for efficient knot free liner changes
  • Proprietary design constructed with the highest quality material and a UV inhibitor ensures long life
  • Rounded handles make lifting and moving easier, and are reinforced to resist tearing or damage from even the heaviest loads
  • Reinforced base is specifically engineered to be dragged over rough surfaces, extending life and overall durability even in the toughest environments
  • Contoured base handles improve grip and ergonomics, reducing strain and improving efficiency when emptying containers
  • Rim features integrated rib structure to increase strength and resist crushing
Weight: 2,7 kg
Certificazioni: Meets NSF 2, 21 approval and is USDA Meat and Poultry Group listed, ensuring regulatory compliance for food storage and clean ability.