Stampa…
Ultra Adhesive Labels
Fornitore: Sampling Systems
8220A-01 8220A-02 8220A-03 8220A-04 8220A-05 8220A-06 8220A-001 8220A-002 8220A-003 8220A-004 8220A-005 8220A-006 8220A-30 8220A-31 8220A-32 8220A-33 8220A-34 8220A-35
300-0550EA 167 EUR
300-0550 SAMP8220A-02 SAMP8220A-03 300-0551 SAMP8220A-05 SAMP8220A-06 300-0548 SAMP8220A-002 SAMP8220A-003 300-0549 SAMP8220A-005 SAMP8220A-006 SAMP8220A-30 300-0552 SAMP8220A-32 SAMP8220A-33 SAMP8220A-34 SAMP8220A-35
Ultra Adhesive Labels
Etichette
Ultra Adhesive (UA) Labels are special high quality labels with an ultra-adhesive foil backing.
  • High strength adhesive - will even stick on very dusty surfaces
  • Working temperature range –20 to +70 °C
  • Dust and weatherproof
UA Labels are ideal for sealing sacks after sampling. The label will not only record the fact that the sack has been sampled but it will prevent the sack from leaking powder as it is handled.