Allyltriphenylphosphonium chloride ≥99%
Fornitore: Thermo Scientific
|
Attenzione
|
A16012.14
A16012.22
A16012.36
A16012.14EA
43.7
EUR
A16012.14
A16012.22
A16012.36
Allyltriphenylphosphonium chloride ≥99%
Allyltriphenylphosphonium chloride
Purity: 99%
CAS No.: 18480-23-4
Molecular Formula: C21H20ClP
Formula Weight: 338.82
Melting Point: 230-232°
MDL No.: MFCD00031542
Formula:
C₂₁H₂₀ClP Peso molecolare: 338,82 g/mol Punto di fusione: 230…232 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00031542 CAS Number: 18480-23-4 EINECS: 242-368-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...