Stampa…

Allyltriphenylphosphonium bromide 99%

Fornitore: Thermo Fisher Scientific
Attenzione

103040250 103041000
ACRO103040250EA 25.2 EUR
ACRO103040250 ACRO103041000
Allyltriphenylphosphonium bromide 99%
Allyltriphenylphosphonium bromide
Formula: C₂₁H₂₀BrP
Punto di fusione: 222…225 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011808
CAS Number: 1560-54-9
EINECS: 216-332-6

Visualizza i codici

Risultati del test di specificazione

Appearance White Powder
Infrared spectrum Conforms
Titration Argentometric ≥98.5 %
Water ≤12 % (K.F.)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR