Stampa…

α-Methylcinnamaldehyde predominantly E isomer 97%

Fornitore: Thermo Scientific
Attenzione

Synonyms: α-Methyl-cinnamaldehyde(predominantly trans)

A10429.22 A10429.36
A10429.22EA 30.8 EUR
A10429.22 A10429.36
α-Methylcinnamaldehyde predominantly E isomer 97%
α-Methylcinnamaldehyde

Purity: 97% CAS No.: 101-39-3 Molecular Formula: C10H10O Formula Weight: 146.19 Flash Point: 79°(174°F) Refractive Index: 1.6050 Boiling Point: 138-140°/14mm Beilstein Reference: 507514 MDL No.: MFCD00006976

Formula: C₁₀H₁₀O
Peso molecolare: 146,19 g/mol
Punto di ebollizione: 138…140 °C (14 mmHg)
Densità: 1,050
Flash Pt: 79 °C (174 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00006976
CAS Number: 101-39-3
EINECS: 202-938-8

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR