Hexamethylphosphoramide ≥98.0% (by GC, titration analysis)
Fornitore: TCI EUROPE
|
Pericolo
|
H0095-25G
H0095-500G
H0095-100G
TCIAH0095-25GEA
19
EUR
TCIAH0095-25G
TCIAH0095-500G
TCIAH0095-100G
Hexamethylphosphoramide ≥98.0% (by GC, titration analysis)
Hexamethylphosphoramide
Formula:
C₆H₁₈N₃OP Punto di ebollizione: 232°C Flash Pt: 132°C |
MDL Number:
MFCD00008303 CAS Number: 680-31-9 EINECS: 211-653-8 Merck Index: 14,04725 |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...