Stampa…

Acetonyltriphenylphosphonium chloride ≥99%

Fornitore: Thermo Fisher Scientific
Attenzione

Synonyms: (2-Oxopropyl)triphenylphosphonium chloride

A14691.14 A14691.22 A14691.36
A14691.14EA 30.9 EUR
A14691.14 A14691.22 A14691.36
Acetonyltriphenylphosphonium chloride ≥99%
Acetonyltriphenylphosphonium chloride

Purity: 99% CAS No.: 1235-21-8 Molecular Formula: C21H20ClOP Formula Weight: 354.82 Melting Point: 244-248° MDL No.: MFCD00011813

Formula: C₂₁H₂₀ClOP
Punto di fusione: 244…248 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011813
CAS Number: 1235-21-8
EINECS: 214-974-1

Visualizza i codici


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR