Stampa…
(Ethoxycarbonylmethyl)triphenylphosphonium bromide ≥98%
Fornitore: Thermo Fisher Scientific
Attenzione
Synonyms: (Carbethoxymethyl)triphenylphosphonium bromide, (2-Ethoxy-2-oxoethyl)triphenylphosphonium bromide
A16347.14 A16347.22
A16347.14EA 37.8 EUR
A16347.14 A16347.22
(Ethoxycarbonylmethyl)triphenylphosphonium bromide ≥98%
(Ethoxycarbonylmethyl)triphenylphosphonium bromide
Purity: 98+% CAS No.: 1530-45-6 Molecular Formula: CH3CH2O2CCH2P(C6H5)3Br Formula Weight: 429.30 Melting Point: ca 150° dec. Beilstein Reference: 3581273
Formula: C₂₂H₂₂BrO₂P
Peso molecolare: 429.30 g/mol
Punto di fusione: ∼ 150 °C (dec.)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00011835
CAS Number: 1530-45-6
EINECS: 216-230-1