Chloromethanesulphonyl chloride ≥98%
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Purity: 98%
CAS No.: 3518-65-8
Molecular Formula: CH2Cl2O2S
Formula Weight: 149.0
Physical Form: Liquid
Density: 1.67
Appearance: Colourless to yellow
Odour: Acidic
Boiling Point: 78-81°/20mm
Beilstein Reference: 1811761
UN No.: 3265
MDL No.: MFCD00014727
Formula:
CH₂Cl₂O₂S Punto di ebollizione: 78…81 °C (20 mmHg) Densità: 1,67 Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00014727 CAS Number: 3518-65-8 EINECS: 222-526-1 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...