Stampa…

Ethyl tiglate ≥98%

Fornitore: Thermo Scientific
Attenzione

A12029.14 A12029.22
A12029.14EA 40.8 EUR
A12029.14 A12029.22
Ethyl tiglate ≥98%
Ethyl tiglate

Purity: 98% CAS No.: 5837-78-5 Molecular Formula: C7H12O2 Formula Weight: 128.17 Flash Point: 46°(114°F) Refractive Index: 1.4340 Boiling Point: 156° Beilstein Reference: 1720895 Merck Reference: 14,9433 UN No.: 3272 MDL No.: MFCD00015183

Formula: C₇H₁₂O₂
Peso molecolare: 128,17 g/mol
Punto di ebollizione: 156 °C
Densità: 0,930
Flash Pt: 46 °C (114 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00015183
CAS Number: 5837-78-5
EINECS: 227-425-6
UN: 3272
ADR: 3,III
Merck Index: 14,9433

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR