8-Aminooctan-1-ol ≥98.0% (by GC, titration analysis)
Fornitore: TCI
|
Attenzione
|
A1522-1G
A1522-5G
TCIAA1522-1GEA
94
EUR
TCIAA1522-1G
TCIAA1522-5G
8-Aminooctan-1-ol ≥98.0% (by GC, titration analysis)
8-Aminooctan-1-ol
Formula:
C₈H₁₉NO Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD02094315 CAS Number: 19008-71-0 |
|
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 98.0 % |
Purity (Nonaqueous Titration) | min. 98.0 % |
Melting point | 60.0 to 65.0 deg-C |
Solubility in Methanol | very faint turbidity |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...