Lanthanum, compound with nickel (1:5) ≥99.5% (metals basis), in polvere -100 mesh
Fornitore: Thermo Fisher Scientific
Synonyms:
Lanthanum nickel
Purity: 99.5%
CAS No.: 12196-72-4
Molecular Formula: LaNi5
Physical Form: Intermetallic, Packaged under argon
Sensitivity: Air and Moisture Sensitive
Odour: Odourless
UN No.: 3178
MDL No.: MFCD00198177
Formula:
LaNi₅ Peso molecolare: 432.36 g/mol |
MDL Number:
MFCD00198177 CAS Number: 12196-72-4 |