Stampa…
Adapters, expansion
Fornitore: witeg Labortechnik
0.510.003 0.510.000 0.510.019
WITG0.510.003EA 15.2 EUR
WITG0.510.003 WITG0.510.000 WITG0.510.019
Adapters, expansion
Adattatori Adattatori conici a smerigliatura
Borosilicate glass 3.3
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Expansion adapters.
Certificazioni: According to DIN 12257.