Stampa…

trans-Cinnamoyl chloride ≥97%

Fornitore: Thermo Scientific
Pericolo

Synonyms: trans-3-Phenylacryloyl Chloride, (E)-Cinnamoyl chloride

A12016.0E A12016.22 A12016.36
A12016.0EEA 855 EUR
A12016.0E A12016.22 A12016.36
trans-Cinnamoyl chloride ≥97%
trans-Cinnamoyl chloride

Purity: 97% CAS No.: 17082-09-6 Molecular Formula: C9H7ClO Formula Weight: 166.61 Flash Point: 110°(230°F) Refractive Index: 1.5845 Odour: Pungent Melting Point: 35-37° Boiling Point: 256-258° Beilstein Reference: 606265 Merck Reference: 14,2301 UN No.: 3261 MDL No.: MFCD00000732

Formula: C₉H₇ClO
Peso molecolare: 166,61 g/mol
Punto di ebollizione: 256…258 °C
Punto di fusione: 35…37 °C
Densità: 1,162
Flash Pt: >110 °C (230 °F)
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00000732
CAS Number: 17082-09-6
EINECS: 203-065-5
UN: 3261
ADR: 8,II
Merck Index: 14,2301

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR