Stampa…
Gilets, Fristads® PR54-521 Design B, marine blue
Fornitore: FRISTADS KANSAS
8515391312XL 851539131L 851539131M 851539131S 851539131XL 851539131XS 8515392102XL 851539210L 851539210M 851539210S 851539210XL 851539210XS 8515392312XL 851539231L 851539231M 851539231S 851539231XL 851539231XS 8515393312XL 851539331L 851539331M 851539331S 851539331XL 851539331XS 8515394402XL 851539440L 851539440M 851539440S 851539440XL 851539440XS 8515395202XL 851539520L 851539520M 851539520S 851539520XL 851539520XS 8515395302XL 851539530L 851539530M 851539530S 851539530XL 851539530XS 8515395392XL 851539539L 851539539M 851539539S 851539539XL 851539539XS 8515396402XL 851539640L 851539640M 851539640S 851539640XL 851539640XS 8515397302XL 851539730L 851539730M 851539730S 851539730XL 851539730XS 8515399402XL 851539940L 851539940M 851539940S 851539940XL 851539940XS 8515399412XL 851539941L 851539941M 851539941S 851539941XL 851539941XS
113-5262EA 0 EUR
113-5262 113-03711 113-03712 113-03713 113-5261 113-03714 113-5258 113-03703 113-03704 113-03705 113-5257 113-03706 113-5264 113-03715 113-03716 113-03717 113-5263 113-03718 113-5266 113-03719 113-03720 113-03721 113-5265 113-03722 113-5268 113-03723 113-03724 113-03725 113-5267 113-03726 113-5248 113-03683 113-03684 113-03685 113-5247 113-03686 113-5252 113-03691 113-03692 113-03693 113-5251 113-03694 113-5250 113-03687 113-03688 113-03689 113-5249 113-03690 113-5254 113-03695 113-03696 113-03697 113-5253 113-03698 113-5256 113-03699 113-03700 113-03701 113-5255 113-03702 113-5270 113-03727 113-03728 113-03729 113-5269 113-03730 113-5260 113-03707 113-03708 113-03709 113-5259 113-03710
Gilets, Fristads® PR54-521 Design B, marine blue
Abbigliamento da lavoro
Gilets made from 65% polyester and 35% cotton; napped inner.
  • Zip fastening
  • 2 breast pockets with flap; front pockets; inner pocket
  • Back pleats; waistband adjustable using press studs, plastic press studs
  • Colour contrasting: Upper collar, bottom edge of zip cover and pocket edges
Material weight: 260 g/m²
Basic colour: Marine
Visualizza i codici
Specifiche
Modello PR54-521, Design B, Marine
Applicazioni per abiti General Purpose
Environmentally Preferable
Risk area
Genere Per l'uomo
Sleeve or leg length Short
Tipo di chiusura Con chiusura a zip
Tessuto 65% polyester and 35% cotton
Peso 260 g/m²