tert-Butyl isocyanide ≥95.0% (by GC)
Fornitore: TCI
|
Pericolo
|
Formula:
C₅H₉N Punto di ebollizione: 92°C Flash Pt: -2°C |
MDL Number:
MFCD00000002 CAS Number: 7188-38-7 EINECS: 230-553-5 UN: 1992 ADR: 3,II |
|
Risultati del test di specificazione
Appearance | Colorless to Almost colorless clear liquid |
Purity (GC) | min. 95.0 % |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...