Stampa…
(S)-(-)-1,1'-Bi-2-naphthol
Fornitore: Cayman Chemical
Synonyms: (S)-BINOL, (S)-(-)-1,1'-Binaphthyl-2,2'-Diol, (S)-[1,1'-Binaphthalene]-2,2'-diol
70052-1 70052-5
CAYM70052-1EA 57.1 EUR
CAYM70052-1 CAYM70052-5
(S)-(-)-1,1'-Bi-2-naphthol
(S)-(-)-1,1'-Bi-2-naphthol
Formula: C₂₀H₁₄O₂
Peso molecolare: 286.33 g/mol
MDL Number: MFCD00004068
CAS Number: 18531-99-2
UN: 2811
ADR: 6.1 III