Terpinolene (p-mentha-1,4(8)-diene) ≥90.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
Terpinolene
T0817-500ML
T0817-25ML
TCIAT0817-500MLEA
58
EUR
TCIAT0817-500ML
TCIAT0817-25ML
Terpinolene (p-mentha-1,4(8)-diene) ≥90.0% (by GC)
Terpinolene (p-mentha-1,4(8)-diene)
Formula:
C₁₀H₁₆ Punto di ebollizione: 185°C Flash Pt: 37°C |
MDL Number:
MFCD00049191 CAS Number: 586-62-9 EINECS: 209-578-0 UN: 2541 ADR: 3,III |
Risultati del test di specificazione
Appearance | Colorless to Light yellow to Light orange clear liquid |
Purity (GC) | min. 90.0 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...