Abietic acid ≥90%
Fornitore: Thermo Scientific
|
Attenzione
|
Purity: 90+%
CAS No.: 514-10-3
Molecular Formula: C20H30O2
Formula Weight: 302.45
Storage Temperature: 2°C to 8°C
Physical Form: Crystalline
Sensitivity: Air Sensitive
Appearance: Pale yellow
Odour: Characteristic
Melting Point: 148-152°
Beilstein Reference: 2221451
Merck Reference: 14,7
MDL No.: MFCD03423567
Formula:
C₂₀H₃₀O₂ Peso molecolare: 302,45 g/mol Punto di fusione: 148…152 °C |
MDL Number:
MFCD03423567 CAS Number: 514-10-3 EINECS: 208-178-3 Merck Index: 14,7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...