Dimcarb (dimethylammonium dimethylcarbamate) (Carbon dioxide:Dimethylamine) <2:1
Fornitore: Thermo Fisher Scientific
|
Pericolo
|
Formula:
(CH₃)₂NH · (CH₃)₂NCOOH Punto di ebollizione: 60…61 °C (760 mmHg) Densità: 1,05 g/cm³ Flash Pt: 13 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00029888 CAS Number: 4137-10-4 UN: 2733 ADR: 3,II |
Risultati del test di specificazione
Appearance | Clear colorless to light yellow Liquid |
Infrared spectrum | Conforms |
Refractive index | 1.4520 to 1.4560 (20°C, 589 nm) |
Water | ≤1 % |
Ratio | < 2.0/1 Carbon dioxide/Dimethylamine |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...