4-Pentylbenzaldehyde 95%
Fornitore: Thermo Fisher Scientific
Purity: 95%
CAS No.: 6853-57-2
Molecular Formula: C12H16O
Formula Weight: 176.26
Refractive Index: 1.5215
Boiling Point: 142-144°/2mm
MDL No.: MFCD00043599
Formula:
C₁₂H₁₆O Punto di ebollizione: 142…144 °C (2 mmHg) Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00043599 CAS Number: 6853-57-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...