5-Chloro-2-methoxybenzaldehyde 97%
Fornitore: Thermo Scientific
|
Attenzione
|
H31860.06
H31860.14
H31860.06EA
81.3
EUR
H31860.06
H31860.14
5-Chloro-2-methoxybenzaldehyde 97%
5-Chloro-2-methoxybenzaldehyde
Purity: 97%
CAS No.: 7035-09-8
Molecular Formula: C8H7ClO2
Formula Weight: 170.60
Odour: Characteristic
Melting Point: 75-79°
MDL No.: MFCD00033854
Formula:
C₈H₇ClO₂ Peso molecolare: 170,60 g/mol Punto di fusione: 75…79 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00033854 CAS Number: 7035-09-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...