Stampa…

(S)-(+)-Methyl-3-hydroxybutyrate ≥98.0% (by GC)

Fornitore: TCI
Pericolo

H0704-25ML H0704-5ML
TCIAH0704-25MLEA 114 EUR
TCIAH0704-25ML TCIAH0704-5ML
(S)-(+)-Methyl-3-hydroxybutyrate ≥98.0% (by GC)
(S)-(+)-Methyl-3-hydroxybutyrate
Formula: C₅H₁₀O₃
Punto di ebollizione: 76°C
Flash Pt: 73°C
MDL Number: MFCD00064461
CAS Number: 53562-86-0
EINECS: 258-628-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f3aade76716b0640ab6d420c25c83081ebb34a76&vid=f3aade76716b0640ab6d420c25c83081ebb34a76&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Colorless to Almost colorless clear liquid
Purity (GC) min. 98.0 %
Specific rotation [a]20/D +21.0 to +24.0 deg (neat)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR