Stampa…

4-tert-Butyl-2-nitrophenol ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: 4-(tert-Butyl)-2-nitrophenol

B1812-25G B1812-5G
TCIAB1812-25GEA 184 EUR
TCIAB1812-25G TCIAB1812-5G
4-tert-Butyl-2-nitrophenol ≥98.0% (by GC)
4-tert-Butyl-2-nitrophenol
Formula: C₁₀H₁₃NO₃
Peso molecolare: 195,2151 g/mol
MDL Number: MFCD00191888
CAS Number: 3279-07-0
EINECS: 221-914-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=403e15f61f76bd2c7280ecc991b491b8a5bc71ba&vid=403e15f61f76bd2c7280ecc991b491b8a5bc71ba&prid=web&clid=SAPDAM

Visualizza i codici

Risultati del test di specificazione

Appearance Orange to Brown clear liquid
Purity (GC) min. 98.0 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR