4-tert-Butylcalix[8]arene 96%
Fornitore: Thermo Fisher Scientific
|
Attenzione
|
Purity: 96%
CAS No.: 68971-82-4
Molecular Formula: C88H112O8
Formula Weight: 1297.87
Melting Point: 411-412°
Beilstein Reference: 2612052
Formula:
C₈₈H₁₁₂O₈ Punto di fusione: 411…412 °C Temperatura di stoccaggio: Ambiente |
MDL Number:
MFCD00075392 CAS Number: 68971-82-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...