(R)-(-)-4-Phenyl-2-oxazolidinone ≥99.0% (by GC)
Fornitore: TCI EUROPE
|
Attenzione
|
Synonyms:
(R)-4-Phenyloxazolidin-2-one
P1307-25G
P1307-5G
TCIAP1307-25GEA
160
EUR
TCIAP1307-25G
TCIAP1307-5G
(R)-(-)-4-Phenyl-2-oxazolidinone ≥99.0% (by GC)
(R)-(-)-4-Phenyl-2-oxazolidinone
Formula:
C₉H₉NO₂ Peso molecolare: 163,18 g/mol |
MDL Number:
MFCD00192393 CAS Number: 90319-52-1 |
Risultati del test di specificazione
Appearance | White to Almost white powder to crystal |
Purity (GC) | min. 99.0 % |
Optical purity (LC) | min. 98.0 ee% |
Melting point | 131.0 to 133.0 deg-C |
Specific rotation [a]20/D | -70.0 to -74.0 deg (C=1, EtOAc) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...