Stampa…

(S)-(-)-4-Benzyl-2-oxazolidinone ≥98.0% (by GC)

Fornitore: TCI EUROPE
Attenzione

Synonyms: (S)-4-Benzyl-2-oxazolidinone, (S)-4-Benzyloxazolidin-2-one

B1754-25G B1754-5G
TCIAB1754-25GEA 53 EUR
TCIAB1754-25G TCIAB1754-5G
(S)-(-)-4-Benzyl-2-oxazolidinone ≥98.0% (by GC)
(S)-(-)-4-Benzyl-2-oxazolidinone
Formula: C₁₀H₁₁NO₂
Peso molecolare: 177,2 g/mol
MDL Number: MFCD00064496
CAS Number: 90719-32-7

Visualizza i codici

Risultati del test di specificazione

Appearance White to Light yellow powder to crystal
Purity (GC) min. 98.0 %
Optical purity (LC) min. 98.0 ee%
Melting point 88.0 to 91.0 deg-C
Specific rotation [a]20/D -14.0 to -15.0 deg (C=5, MeOH)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR