Stampa…
Titanium diboride
Fornitore: Thermo Scientific
Synonyms: Titanium boride
11364.18 11364.30 11364.A1
11364.18EA 53.6 EUR
11364.18 11364.30 11364.A1
Titanium diboride
Titanium diboride
CAS No.: 12045-63-5 Molecular Formula: B2Ti Formula Weight: 69.50 Physical Form: -325 Mesh Powder Density: 4.38 Odour: Odourless MDL No.: MFCD00049595
In the aircraft industry, in cutting and grinding applications, as protective coatings for metals in high temperature applications, as a base material for cermet production
Formula: B₂Ti
Peso molecolare: 69,5 g/mol
Punto di fusione: 3225 °C
Densità: 4,38
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD00049595
CAS Number: 12045-63-5
EINECS: 234-961-4