Monoethyl fumarate calcium salt per sintesi
Fornitore: Merck
Formula:
C₁₂H₁₄CaO₈ |
CAS Number:
62008-22-4 EINECS: 263-369-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...