Stampa…

L(-)-Lactamide per sintesi, Sigma-Aldrich®

Fornitore: Merck


M-Clarity™ quality level = MQ200

8.20752.1000 8.20752.9014 8.20752.9022 8.20752.9023
8.20752.1000EA 0 EUR
8.20752.1000 8.20752.9014 8.20752.9022 8.20752.9023
L(-)-Lactamide per sintesi, Sigma-Aldrich®
L(-)-Lactamide
Formula: C₃H₇NO₂
MDL Number: MFCD00198010
CAS Number: 89673-71-2

Visualizza i codici

Risultati del test di specificazione

Solubility 782 g/l (25°C) (experimental)
Melting Point 50 - 53 °C
Molar Mass 89.09 g/mol


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR