Stampa…

6-Fluoronicotinic acid ≥97%

Fornitore: Thermo Scientific
Attenzione

Synonyms: 2-Fluoro-5-pyridinecarboxylic acid

H55147.MD H55147.03
H55147.MDEA 90.6 EUR
H55147.MD H55147.03
6-Fluoronicotinic acid ≥97%
6-Fluoronicotinic acid

Purity: 97% CAS No.: 403-45-2 Molecular Formula: C6H4FNO2 Formula Weight: 141.10 Melting Point: 144-148° MDL No.: MFCD01859863

Formula: C₆H₄FNO₂
Peso molecolare: 141,1 g/mol
Punto di fusione: 144…148 °C
Temperatura di stoccaggio: Ambiente
MDL Number: MFCD01859863
CAS Number: 403-45-2

Visualizza i codici

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Informazioni About VWR